KEY293014915 Key Organics / BIONET

[5-(trifluoromethyl)pyridin-2-yl]methanol

  • MDL number MFCD04117762
  • Cas Number : 31181-84-7
  • Molecular Weight : 177.126

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 76 - 78 (1mm Hg)
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96480

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EE-0713_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart