KEY293014916 Key Organics / BIONET

1-(diphenylmethyl)-3-phenylazetidine

  • MDL number MFCD16622779
  • Cas Number : 913814-30-9
  • Molecular Weight : 299.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 77 - 78
Iupac Name 1-benzhydryl-3-phenylazetidine
Inchi Code 1S/C22H21N/c1-4-10-18(11-5-1)21-16-23(17-21)22(19-12-6-2-7-13-19)20-14-8-3-9-15-20/h1-15,21-22H,16-17H2
Inchi Key PAXDHGAELYIMBD-UHFFFAOYSA-N

Description

Melting Point 77 - 78
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96170

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DE-0211_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart