KEY293014923 Key Organics / BIONET

1-(4-methylbenzenesulfonyl)-2,3,4,5-tetrahydro-1H-1-benzazepin-5-one

  • MDL number MFCD00795994
  • Cas Number : 24310-36-9
  • Molecular Weight : 315.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 125
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94660

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AE-0248_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart