KEY293014926 Key Organics / BIONET

3,5-dibromo-2-chloropyridin-4-amine

  • MDL number MFCD11110699
  • Cas Number : 1054484-40-0
  • Molecular Weight : 286.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 156 - 157
Iupac Name 3,5-dibromo-2-chloro-4-pyridinamine
Inchi Code 1S/C5H3Br2ClN2/c6-2-1-10-5(8)3(7)4(2)9/h1H,(H2,9,10)
Inchi Key JSWRREFUDHGVFW-UHFFFAOYSA-N

Description

Melting Point 156 - 157
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96494

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/EE-0741_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart