KEY298198372 Key Organics / BIONET

1-tert-butyl 2-methyl (2S,4R)-4-hydroxypiperidine-1,2-dicarboxylate

  • MDL number MFCD08669704
  • Cas Number : 181269-87-4
  • Molecular Weight : 259.302

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88 - 90
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96466

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EE-0204_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart