KEY298198421 Key Organics / BIONET

2-{[(tert-butoxy)carbonyl]amino}-4,5-dimethoxybenzoic acid

  • MDL number MFCD02682186
  • Cas Number : 122744-78-9
  • Molecular Weight : 297.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[(tert-butoxycarbonyl)amino]-4,5-dimethoxybenzoic acid
Inchi Code 1S/C14H19NO6/c1-14(2,3)21-13(18)15-9-7-11(20-5)10(19-4)6-8(9)12(16)17/h6-7H,1-5H3,(H,15,18)(H,16,17)
Inchi Key ZXNOFBMUVBIGOE-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108534

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4051_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart