KEY298198427 Key Organics / BIONET

tert-butyl piperidine-3-carboxylate hydrochloride

  • MDL number MFCD04038695
  • Cas Number : 301180-05-2
  • Molecular Weight : 221.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-piperidinecarboxylate hydrochloride
Inchi Code 1S/C10H19NO2.ClH/c1-10(2,3)13-9(12)8-5-4-6-11-7-8;/h8,11H,4-7H2,1-3H3;1H
Inchi Key FNTUZXRGYYMCBP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108543

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4071_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart