KEY298198435 Key Organics / BIONET

1-(piperidin-4-yl)-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD02178923
  • Cas Number : 1187174-05-5
  • Molecular Weight : 237.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108556

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4101_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart