KEY298198504 Key Organics / BIONET

3-chloro-N2-methylpyridine-2,5-diamine

  • MDL number MFCD16622774
  • Cas Number : 1228666-61-2
  • Molecular Weight : 157.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point LMS
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95772

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CE-0739_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart