KEY298198509 Key Organics / BIONET

4-{3-chloro-5-[(4-cyanophenyl)amino]phenoxy}-3,5-dimethylbenzonitrile

  • MDL number MFCD16710267
  • Cas Number : 1242268-27-4
  • Molecular Weight : 373.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 278 - 280
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96938

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FE-0066_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart