KEY298198527 Key Organics / BIONET

3,4-dihydro-2H-1,4-benzoxazin-6-amine hydrochloride

  • MDL number MFCD16710277
  • Cas Number : 207596-59-6
  • Molecular Weight : 186.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 250
Purity 95
Storage temp. RT
Physical Form Grey to Black Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97009

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FE-0777_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart