KEY298198549 Key Organics / BIONET

ethyl 1H-1,3-benzodiazole-2-carboxylate

  • MDL number MFCD00721392
  • Cas Number : 1865-09-4
  • Molecular Weight : 190.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point ~ 220
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96981

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FE-0733_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart