KEY298198550 Key Organics / BIONET

tert-butyl 2-(chloromethyl)-1H-1,3-benzodiazole-1-carboxylate

  • MDL number MFCD10697638
  • Cas Number : 163798-87-6
  • Molecular Weight : 266.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80 - 82
Purity 97
Storage temp. RT
Physical Form White to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96860

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FD-0036_SDS.pdf
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart