KEY298198553 Key Organics / BIONET

4-amino-3-methoxybenzonitrile

  • MDL number MFCD13250177
  • Cas Number : 177476-76-5
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point LMS
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96986

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/FE-0741_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart