KEY298198558 Key Organics / BIONET

5-chloro-2-(chloromethyl)pyridine

  • MDL number MFCD10697588
  • Cas Number : 10177-24-9
  • Molecular Weight : 162.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Storage temp. Store under N2 at 4�Light sensitive
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96991

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P410,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FE-0751_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart