KEY298198568 Key Organics / BIONET

methyl 3-hydroxy-4-iodobenzoate

  • MDL number MFCD06203960
  • Cas Number : 157942-12-6
  • Molecular Weight : 278.045

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 167
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97005

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FE-0770_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart