KEY298198580 Key Organics / BIONET

2-(propan-2-yl)piperazine

  • MDL number MFCD07373042
  • Cas Number : 84468-53-1
  • Molecular Weight : 128.219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point LMS
Purity 95
Storage temp. Store under N2, Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91373

Safety Information

signalword Warning
pcodes P232,P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Z-0600_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart