KEY298198581 Key Organics / BIONET

7-methoxy-4H,5H-pyrrolo[1,2-a]quinoxalin-4-one

  • MDL number MFCD16710282
  • Cas Number : 160657-07-8
  • Molecular Weight : 214.224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 249 - 251
Purity 95
Country of Origin United Kingdom
Product Link http://www.keyorganics.net/bionet/catalog/product/view/id/97463

Safety Information

signalword Warning
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart