KEY298198583 Key Organics / BIONET

ethyl 5-bromo-2-chloropyridine-4-carboxylate

  • MDL number MFCD14698168
  • Cas Number : 1214346-11-8
  • Molecular Weight : 264.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp Oil
Iupac Name ethyl 5-bromo-2-chloroisonicotinate
Inchi Code 1S/C8H7BrClNO2/c1-2-13-8(12)5-3-7(10)11-4-6(5)9/h3-4H,2H2,1H3
Inchi Key YVGLNIDGZXOBSZ-UHFFFAOYSA-N

Description

Melting Point Oil
Purity 95%
Physical Form Liquid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96957

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FE-0247_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart