KEY298198589 Key Organics / BIONET

4-tert-butyl 2-methyl (2R,5S)-2-(2-methylpropyl)-5-(1,3-thiazol-2-yl)pyrrolidine-2,4-dicarboxylate

  • MDL number MFCD12025840
  • Cas Number : 1820598-57-9
  • Molecular Weight : 368.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94529

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AD-0323_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart