KEY298198592 Key Organics / BIONET

{1H-pyrrolo[2,3-b]pyridin-3-yl}methanol

  • MDL number MFCD11054203
  • Cas Number : 1065100-83-5
  • Molecular Weight : 148.165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 171 - 173
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97481

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GE-0726_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart