KEY298198600 Key Organics / BIONET

tert-butyl 3-(hydroxymethyl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

  • MDL number MFCD11054204
  • Cas Number : 144657-67-0
  • Molecular Weight : 248.282

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146 - 148
Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97490

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GE-0740_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart