KEY298198603 Key Organics / BIONET

(1-methyl-1H-1,2,4-triazol-3-yl)methanol

  • MDL number MFCD16710296
  • Cas Number : 135242-93-2
  • Molecular Weight : 113.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72 - 75
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97494

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GE-0745_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart