KEY298198604 Key Organics / BIONET

3-(chloromethyl)-1-methyl-1H-1,2,4-triazole hydrochloride

  • MDL number MFCD09701417
  • Cas Number : 135206-76-7
  • Molecular Weight : 168.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69 - 70
Purity 95
Storage temp. Store under N2
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97495

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
hcodes H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/GE-0746_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart