KEY298198607 Key Organics / BIONET

6-hydroxy-2-phenylpyrimidine-4-carboxylic acid

  • MDL number MFCD00856151
  • Cas Number : 84659-98-3
  • Molecular Weight : 216.196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 265 - 267
Purity 95
Storage temp. RT
Physical Form White to Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101807

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LD-0088_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart