KEY298198610 Key Organics / BIONET

4-chloro-2,6-bis(trifluoromethyl)pyridine

  • MDL number MFCD00082558
  • Cas Number : 81269-96-7
  • Molecular Weight : 249.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57 - 59
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3710

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/10Z-7005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart