KEY298198615 Key Organics / BIONET

3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}butanoic acid

  • MDL number MFCD00270340
  • Cas Number : 186320-18-3
  • Molecular Weight : 325.364

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108533

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4050_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart