KEY299910980 Key Organics / BIONET

2-methyl-2H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD00053366
  • Cas Number : 42150-25-4
  • Molecular Weight : 116.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108569

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4159_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart