KEY299911027 Key Organics / BIONET

pyrimidine-2-carboximidamide

  • MDL number MFCD09755980
  • Cas Number : 45695-56-5
  • Molecular Weight : 122.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 162 - 164
Iupac Name 2-pyrimidinecarboximidamide
Inchi Code 1S/C5H6N4/c6-4(7)5-8-2-1-3-9-5/h1-3H,(H3,6,7)
Inchi Key SZSKAHAHBFDQKN-UHFFFAOYSA-N

Description

Melting Point 162 - 164
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97454

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GE-0236_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart