KEY299911039 Key Organics / BIONET

1-(2,2-dimethoxyethyl)-2-nitrobenzene

  • MDL number MFCD16890134
  • Cas Number : 79844-33-0
  • Molecular Weight : 211.217

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97822

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/HE-0029_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart