KEY299911040 Key Organics / BIONET

1-(4-iodophenyl)piperidin-2-one

  • MDL number MFCD04037128
  • Cas Number : 385425-15-0
  • Molecular Weight : 301.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 118 - 120
Iupac Name 1-(4-iodophenyl)-2-piperidinone
Inchi Code 1S/C11H12INO/c12-9-4-6-10(7-5-9)13-8-2-1-3-11(13)14/h4-7H,1-3,8H2
Inchi Key OYDSTJMVOOOYDW-UHFFFAOYSA-N

Description

Melting Point 118 - 120
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97825

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/HE-0041_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart