KEY309658637 Key Organics / BIONET

2-[3-(methoxycarbonyl)-5-methyl-1H-pyrazol-1-yl]benzoic acid

  • MDL number MFCD17167308
  • Cas Number : 1246738-24-8
  • Molecular Weight : 260.249

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95278

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-0009_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart