KEY309658689 Key Organics / BIONET

7-fluoro-2,3-dihydro-1H-inden-4-ol

  • MDL number MFCD17167331
  • Cas Number : 161178-24-1
  • Molecular Weight : 152.168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108602

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4323_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart