KEY309658839 Key Organics / BIONET

tert-butyl 4-(6-methyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)piperidine-1-carboxylate

  • MDL number MFCD17392914
  • Cas Number : 950772-99-3
  • Molecular Weight : 331.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 194 - 196
Iupac Name tert-butyl 4-(6-methyl-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl)-1-piperidinecarboxylate
Inchi Code 1S/C18H25N3O3/c1-12-5-6-14-15(11-12)21(16(22)19-14)13-7-9-20(10-8-13)17(23)24-18(2,3)4/h5-6,11,13H,7-10H2,1-4H3,(H,19,22)
Inchi Key OAWZQSMENFQQHO-UHFFFAOYSA-N

Description

Melting Point 194 - 196
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97818

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/HE-0020_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart