KEY309658860 Key Organics / BIONET

5-[(4-chlorophenyl)methyl]-1,3,4-oxadiazol-2-ol

  • MDL number MFCD17392925
  • Cas Number : 34547-04-1
  • Molecular Weight : 210.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 112 - 114
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101891

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LE-0717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart