KEY309658879 Key Organics / BIONET

2-(4-chlorophenyl)acetohydrazide

  • MDL number MFCD00157887
  • Cas Number : 57676-51-4
  • Molecular Weight : 184.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 (lit)
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101069

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KE-0780_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart