KEY309658891 Key Organics / BIONET

2-(1-methyl-1H-indol-3-yl)acetamide

  • MDL number MFCD02683891
  • Cas Number : 150114-41-3
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180 - 182
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97449

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GE-0097_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart