KEY309658903 Key Organics / BIONET

2-[2-(2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}acetamido)acetamido]acetic acid

  • MDL number MFCD00190881
  • Cas Number : 170941-79-4
  • Molecular Weight : 411.414

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108573

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4172_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart