KEY309658908 Key Organics / BIONET

9H-fluoren-9-ylmethyl N-(piperidin-3-ylmethyl)carbamate

  • MDL number MFCD03001743
  • Cas Number : 672310-14-4
  • Molecular Weight : 336.435

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108594

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4275_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart