KEY311758405 Key Organics / BIONET

5-hydroxy-4-methoxy-2-nitrobenzoic acid

  • MDL number MFCD10666941
  • Cas Number : 31839-20-0
  • Molecular Weight : 213.145

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178 - 180 (dec)
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94735

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AF-0707_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart