KEY311758408 Key Organics / BIONET

methyl N-[(3S,4S)-4-methylpiperidin-3-yl]carbamate

  • MDL number MFCD09030046
  • Cas Number : 694495-64-2
  • Molecular Weight : 172.228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point LMS
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102254

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/ME-0136_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart