KEY311758466 Key Organics / BIONET

3-(4-chlorophenyl)-4-fluoro-1H-pyrazol-5-amine

  • MDL number MFCD18426511
  • Cas Number : 1246384-94-0
  • Molecular Weight : 211.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163 - 165
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94734

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AF-0706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart