KEY311758471 Key Organics / BIONET

{1-methyl-3-phenyl-1H-thieno[2,3-c]pyrazol-5-yl}methanol

  • MDL number MFCD18426519
  • Cas Number : 1263063-15-5
  • Molecular Weight : 244.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 135 - 137
Iupac Name (1-methyl-3-phenyl-1H-thieno[2,3-c]pyrazol-5-yl)methanol
Inchi Code 1S/C13H12N2OS/c1-15-13-11(7-10(8-16)17-13)12(14-15)9-5-3-2-4-6-9/h2-7,16H,8H2,1H3
Inchi Key ARPBBSGGQDLQED-UHFFFAOYSA-N

Description

Melting Point 135 - 137
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94751

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AF-0727_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart