KEY311758477 Key Organics / BIONET

2-fluoro-3-oxo-3-phenylpropanenitrile

  • MDL number MFCD18426513
  • Cas Number : 343271-85-2
  • Molecular Weight : 163.151

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 24 - 26
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94737

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AF-0709_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart