KEY311758478 Key Organics / BIONET

4-chloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD12924542
  • Cas Number : 582313-57-3
  • Molecular Weight : 171.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 177 - 180 (dec)
Iupac Name 4-chloro-5-fluoro-7H-pyrrolo[2,3-d]pyrimidine
Inchi Code 1S/C6H3ClFN3/c7-5-4-3(8)1-9-6(4)11-2-10-5/h1-2H,(H,9,10,11)
Inchi Key OOBFPXOXHUBRDN-UHFFFAOYSA-N

Description

Melting Point 177 - 180 (dec)
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94741

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/AF-0713_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart