KEY311758614 Key Organics / BIONET

2-fluoro-3-oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile

  • MDL number MFCD18426514
  • Cas Number : 1263063-11-1
  • Molecular Weight : 231.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94738

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AF-0710_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart