KEY311758617 Key Organics / BIONET

4-fluoro-3-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD18426518
  • Molecular Weight : 245.181

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111 - 113
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94750

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AF-0726_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart