KEY311758630 Key Organics / BIONET

1,3-dimethyl-1H,4H,5H,6H,7H-pyrazolo[3,4-d]pyrimidine-4,6-dione

  • MDL number MFCD18157655
  • Cas Number : 1072895-79-4
  • Molecular Weight : 180.167

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 300
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101849

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LD-0727_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart