KEY311758646 Key Organics / BIONET

6-hydroxy-[2,2'-bipyrimidine]-4-carboxylic acid

  • MDL number MFCD18157673
  • Cas Number : 1261079-77-9
  • Molecular Weight : 218.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238 - 241
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/102277

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/ME-0715_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart