KEY311758695 Key Organics / BIONET

1-[4-(3-chloropropoxy)-3-methoxyphenyl]ethan-1-one

  • MDL number MFCD05156572
  • Cas Number : 58113-30-7
  • Molecular Weight : 242.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 61 - 64
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106793

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/NE-0054_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart